Learn More
Cayman Chemical Ticarcillin (sodium salt), Purity: ≥95% (mixture of diastereomers), CAS: 4697-14-7, 1 g
SDP
Catalog No. 502934111
Encompass_Preferred
A semi-synthetic β-lactam antibiotic; active against P. aeruginosa, E. coli, P. mirabilis, P. rettgeri, and K. aerogenes (MICs = 4-125 μg/ml); reduces P. aeruginosa colony count in rabbit eye at 2.5 mg/eye
Catalog No. 50-293-4111 Supplier Cayman Chemical Supplier No. 260651 G
A semi-synthetic β-lactam antibiotic; active against P. aeruginosa, E. coli, P. mirabilis, P. rettgeri, and K. aerogenes (MICs = 4-125 μg/ml); reduces P. aeruginosa colony count in rabbit eye at 2.5 mg/eye