Learn More
Cayman Chemical TIcarcIlInsodIum salt 25g
SDP
Catalog No. 502936319
Encompass_Preferred
A semi-synthetic β-lactam antibiotic; active against P. aeruginosa, E. coli, P. mirabilis, P. rettgeri, and K. aerogenes (MICs = 4-125 μg/ml); reduces P. aeruginosa colony count in rabbit eye at 2.5 mg/eye
Catalog No. 50-293-6319 Supplier Cayman Chemical Supplier No. 2606525 G
A semi-synthetic β-lactam antibiotic; active against P. aeruginosa, E. coli, P. mirabilis, P. rettgeri, and K. aerogenes (MICs = 4-125 μg/ml); reduces P. aeruginosa colony count in rabbit eye at 2.5 mg/eye