Learn More
Gold Biotechnology Inc Timentin™ Ticarcillin/Clavulanate (15/1) 25 g
SDP
Catalog No. 501532840
Encompass_Preferred
Timentin™ is a 15:1 mixture of ticarcillin and clavulanate. Ticarcillin is a penicillin ß-lactam antibiotic, which is susceptible to ß-lactamase degradation. When combined with clavulanate, a ß-lactamase inhibitor, its efficacy is greatly increased. Timentin™ exhibits high activity against gram-negative bacteria and resistant Agrobacterium species.
Catalog No. 50-153-2840 Supplier Gold Biotechnology Inc Supplier No. T10425
Timentin™ is a 15:1 mixture of ticarcillin and clavulanate. Ticarcillin is a penicillin ß-lactam antibiotic, which is susceptible to ß-lactamase degradation. When combined with clavulanate, a ß-lactamase inhibitor, its efficacy is greatly increased. Timentin™ exhibits high activity against gram-negative bacteria and resistant Agrobacterium species.