Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ticarcillin/Clavulanate, G-Biosciences
SCI_ED

Catalog No. S139791
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S139791 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S139791 Supplier G-Biosciences Supplier No. RC872
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High activity against gram-negative bacteria and the combination is effective against resistant Agrobacterium strains.

Ticarcillin is an inhibitor of bacterial cell wall synthesis. It inhibits the crosslinking of peptidoglycan by binding and inactivating of transpeptidases.

Chemical Identifiers

Molecular Weight (g/mol) 665.64

Specifications

Physical Form Powder
Quantity 500 g
Product Line Gibco
Boiling Point 768.3°C
Percent Purity 0.95
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.