Learn More
Alkali Scientific Ticarcillin Disodium Salt, 5 G
SDP
Catalog No. 5000661196
Encompass_Preferred
Ticarcillin Disodium Salt (5g) is an antibiotic used for treating infections caused by a variety of Gram-negative and some Gram-positive bacteria. This powder form is easily reconstituted into a solution for intravenous use in treating severe infections like pneumonia, urinary tract infections, and bloodstream infections. Ticarcillin works by inhibiting bacterial cell wall synthesis, making it effective against fast-growing pathogens such as Pseudomonas aeruginosa. The 5g packaging is ideal for hospitals and laboratories requiring smaller quantities for targeted treatment or research, and it is frequently used in combination with other antibiotics to enhance therapeutic outcomes.
Catalog No. 50-006-61196 Supplier Alkali Scientific Supplier No. TIC002CP
Ticarcillin Disodium Salt (5g) is an antibiotic used for treating infections caused by a variety of Gram-negative and some Gram-positive bacteria. This powder form is easily reconstituted into a solution for intravenous use in treating severe infections like pneumonia, urinary tract infections, and bloodstream infections. Ticarcillin works by inhibiting bacterial cell wall synthesis, making it effective against fast-growing pathogens such as Pseudomonas aeruginosa. The 5g packaging is ideal for hospitals and laboratories requiring smaller quantities for targeted treatment or research, and it is frequently used in combination with other antibiotics to enhance therapeutic outcomes.