Learn More
Research Products International Corp Ticarcillin Disodium Salt, 5 Grams
SDP
Catalog No. 501368646
Encompass_Preferred
Ticarcillin disodium salt is an antibiotic that belongs to the class of drugs known as penicillin. It is a broad-spectrum antibiotic effective against various bacterial infections.
Ticarcillin disodium salt is commonly used in the field of life sciences for research applications.
In the life science marketplace, ticarcillin disodium salt is primarily utilized as a selective agent in microbiology and molecular biology experiments.
It is often incorporated into culture media to inhibit the growth of certain bacteria, allowing for the selective growth of desired organisms or the detection of specific bacterial strains.
Researchers and scientists working in fields such as microbiology, immunology, and molecular biology may employ ticarcillin disodium salt to study bacterial resistance mechanisms, evaluate the efficacy of antimicrobial compounds, or isolate and identify specific bacterial species.
Catalog No. 50-136-8646 Supplier Research Products International Corp Supplier No. T365005.0
Ticarcillin disodium salt is an antibiotic that belongs to the class of drugs known as penicillin. It is a broad-spectrum antibiotic effective against various bacterial infections.
Ticarcillin disodium salt is commonly used in the field of life sciences for research applications.
In the life science marketplace, ticarcillin disodium salt is primarily utilized as a selective agent in microbiology and molecular biology experiments.
It is often incorporated into culture media to inhibit the growth of certain bacteria, allowing for the selective growth of desired organisms or the detection of specific bacterial strains.
Researchers and scientists working in fields such as microbiology, immunology, and molecular biology may employ ticarcillin disodium salt to study bacterial resistance mechanisms, evaluate the efficacy of antimicrobial compounds, or isolate and identify specific bacterial species.