Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ticarcillin Disodium salt, G-Biosciences
SCI_ED

Catalog No. S000018
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S000018 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S000018 Supplier G-Biosciences Supplier No. RC1119
Add to Cart
Add to Cart

Ticarcillin is a carboxypenicillin used to treat gram-negative bacteria like Pseudomonas aeruginosa and Stenotrophomonas maltophilia.

An alternative to ampicillin, it can be used to test the presence of marker genes in bacteria, while preventing the appearance of satellite colonies occurring during the ampicillin breakdown.

Chemical Identifiers

Molecular Weight (g/mol) 428.39 g/mol

Specifications

Quantity 25 g
Percent Purity >80%
Grade Laboratory
Physical Form Powder
Product Line Gibco
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.