Learn More
Bioworld Tick-Borne Encephalitis Virus gE (Recombinant), 100 μg
Supplier:  Bioworld 220607161
Encompass_Preferred
Recombinant Tick-Born Encephalitis Virus glycoprotein E (gE) (95-229) Source: E.coli Description : The protein contains the Tick-borne Encephalitis Virus gE regions, amino acids:(95-229). Purification method: Purified by proprietary chromatographic technique. Purity: Protein is >95% pure as determined by 10% PAGE (coomassie staining). Protein conc: 1mg/ml in 20mM MES pH 6.5, 8M urea, 200 mM NaCL and 0.05% Tween-20. Storage: Protein is shipped at ambient temperature. Upon arrival, Store at -20C. Stability: Five years frozen. One month in solution at room temperature. Specificity: Immunoreactive with sera of encephalitis virus infected individuals. Application: Antigen in ELISA and Western blots, excellent antigen for detection of Tick-borne encephalitis virus with minimal specificity problems. Usage: BioWORLD\\\'s products are furnished for LABORATORY RESEARCH USE ONLY. The product may not be used as drugs, agricultural or pesticidal products, food additives or household chemicals.
Catalog No. 50-253-814