Learn More
Description
- Explore the characteristics of various tick species
- Extract DNA from simulated tick samples
- Use PCR to amplify pathogenic DNA within a species
- Compare PCR band patterns to identify tick borne disease present in a geographic region
- Group size: For 24 reactions, or 6 groups of 4 students
- Kit includes: Instructions, PCR EdvoBeads™ PLUS, Universal DNA Buffer, TE Buffer, LyphoPrimer™ Mix, LyphoControl™ Tick DNA Templates, EdvoQuick™ DNA Ladder, SYBR™ Safe DNA Stain, Ultra-Spec Agarose™, 50x Electrophoresis Buffer, 0.2 mL PCR Tubes, Plastic Micropestles, Screw-Top Tubes, Snap-Top Microcentrifuge Tubes, Simulated Ticks
- All you need: DNA Electrophoresis Apparatus and Power Supply, Thermal Cycler, Micropipettes: 5-50 μl, UV or Blue Light Transilluminator, Microcentrifuge, Microwave or Hot plate, bleach solution, ice buckets and ice, distilled or deionized water, hot gloves, disposable lab gloves, 250 mL flasks or beakers, pipet pump
- Storage: Some Components Require Freezer Storage Upon Receipt
Specifications
Specifications
| Grade Level | High School, Higher Ed |
| Class Size | 6 Lab Groups |
| Product Type | DNA Extraction and Isolation Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

