Learn More
Cayman Chemical Ticlopidine-d4 (hydrochloride), Purity: ≥99% deuterated forms (d1-d4), CAS: 2932627-17-1, 1 mg
SDP
Catalog No. 502948626
Encompass_Preferred
An internal standard for the quantification of ticlopidine by GC- or LC-MS
Catalog No. 50-294-8626 Supplier Cayman Chemical Supplier No. 334671 MG
An internal standard for the quantification of ticlopidine by GC- or LC-MS