Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ticlopidine hydrochloride, 100.87%, MP Biomedicals™

Catalog No. ICN15692205 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
ICN15692205 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15692205 Supplier MP Biomedicals, Inc Supplier No. 0215692205

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

Specifications

CAS 53885-35-1
Color White
Assay Percent Range 1.0087
Molecular Formula C14H15Cl2NS
MDL Number MFCD00079606
Quantity 5 g
Synonym ticlopidine hydrochloride, ticlopidine hcl, ticlodix, ticlodone, ticlid, panaldine, tiklid, 5-2-chlorobenzyl-4,5,6,7-tetrahydrothieno 3,2-c pyridine hydrochloride, unii-a1l4914fmf
InChI Key MTKNGOHFNXIVOS-UHFFFAOYSA-N
SMILES [H+].[Cl-].ClC1=CC=CC=C1CN1CCC2=C(C1)C=CS2
IUPAC Name hydrogen 5-[(2-chlorophenyl)methyl]-4H,5H,6H,7H-thieno[3,2-c]pyridine chloride
Molecular Weight (g/mol) 300.24
PubChem CID 65335
ChEBI CHEBI:9589
Formula Weight 300.2
Physical Form Free-flowing Powder
Chemical Name or Material Ticlopidine hydrochloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.