Learn More
Medchemexpress LLC Ticlopidine hydrochloride | 53885-35-1 | 99.99% | 1 ML
SDP
Catalog No. 5000407236
Encompass_Preferred
Ticlopidine hydrochloride is an antithrombotic proagent that acts as an allosteric, noncompetitive inhibitor of CD39. It also blocks several NTPDase isoenzymes and inhibits human liver cytochromes CYP2C19, CYP2C9, and CYP3A4.
  • Acts as an allosteric, noncompetitive inhibitor of CD39 (IC50 of 81.7 μM)
  • Blocks NTPDase isoenzymes, including NTPDase2 and NTPDase3
  • Inhibits CYP2C19 human liver cytochrome
  • Inhibits CYP2C9 (IC50 of 26.0 μM)
  • Inhibits CYP3A4 (IC50 of 32.3 μM)
Catalog No. 50-004-07236 Supplier Medchemexpress LLC Supplier No. HYB0153A10MM/1ML
Ticlopidine hydrochloride is an antithrombotic proagent that acts as an allosteric, noncompetitive inhibitor of CD39. It also blocks several NTPDase isoenzymes and inhibits human liver cytochromes CYP2C19, CYP2C9, and CYP3A4.
  • Acts as an allosteric, noncompetitive inhibitor of CD39 (IC50 of 81.7 μM)
  • Blocks NTPDase isoenzymes, including NTPDase2 and NTPDase3
  • Inhibits CYP2C19 human liver cytochrome
  • Inhibits CYP2C9 (IC50 of 26.0 μM)
  • Inhibits CYP3A4 (IC50 of 32.3 μM)