Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ticlopidine, Hydrochloride, MP Biomedicals™

Catalog No. ICN15692225 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15692225 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15692225 Supplier MP Biomedicals, Inc Supplier No. 0215692225

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 53885-35-1
Molecular Formula C14H15Cl2NS
Molecular Weight (g/mol) 300.24
MDL Number MFCD00079606
InChI Key MTKNGOHFNXIVOS-UHFFFAOYSA-N
Synonym ticlopidine hydrochloride, ticlopidine hcl, ticlodix, ticlodone, ticlid, panaldine, tiklid, 5-2-chlorobenzyl-4,5,6,7-tetrahydrothieno 3,2-c pyridine hydrochloride, unii-a1l4914fmf
PubChem CID 65335
ChEBI CHEBI:9589
IUPAC Name hydrogen 5-[(2-chlorophenyl)methyl]-4H,5H,6H,7H-thieno[3,2-c]pyridine chloride
SMILES [H+].[Cl-].ClC1=CC=CC=C1CN1CCC2=C(C1)C=CS2

Specifications

Quantity 25 g
Formula Weight 300.2
Physical Form Solid
Chemical Name or Material Ticlopidine, Hydrochloride
Recommended Storage Store at 2° to 8°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.