Learn More
Cayman Chemical Ticlopidine (hydrochloride), Purity: ≥98%, CAS: 53885-35-1, 1 g
SDP
Catalog No. 502984135
Encompass_Preferred
A P2Y12 receptor antagonist; inhibits collagen-, arachidonic acid-, and ADP-induced aggregation of human platelets (IC50s = 75, 600, and 1,300 μM, respectively); inhibits ADP-induced aggregation of rat platelets; decreases thrombus weight in a rat model of arterio-venous shunt thrombosis at 100 mg/kg; inhibits healing of acetic acid-induced gastric ulcers in rats at 300 mg/kg
Catalog No. 50-298-4135 Supplier Cayman Chemical Supplier No. 207701 G
A P2Y12 receptor antagonist; inhibits collagen-, arachidonic acid-, and ADP-induced aggregation of human platelets (IC50s = 75, 600, and 1,300 μM, respectively); inhibits ADP-induced aggregation of rat platelets; decreases thrombus weight in a rat model of arterio-venous shunt thrombosis at 100 mg/kg; inhibits healing of acetic acid-induced gastric ulcers in rats at 300 mg/kg