Learn More
Aat Bioquest Tide Fluor™ 3 maleimide [TF3 maleimide] *Superior replacement for Cy3*
SDP
Catalog No. 501953129
Encompass_Preferred
Tide Fluor™ 3 (TF3) family has the spectral properties essentially identical to those of Cy3. Compared to Cy3 probes TF3 family has much stronger fluorescence and higher photostability.
Catalog No. 50-195-3129 Supplier Aat Bioquest Supplier No. 2270
Tide Fluor™ 3 (TF3) family has the spectral properties essentially identical to those of Cy3. Compared to Cy3 probes TF3 family has much stronger fluorescence and higher photostability.