Learn More
Aat Bioquest Tide Quencher™ 3 maleimide [TQ3 maleimide]
SDP
Catalog No. 501953104
Encompass_Preferred
TQ3 is designed to be a superior quencher to TAMRA, TF3 and Cy3. TQ3 has (a). much stronger absorption; (b). much higher quenching efficiency; and (c). versatile reactive forms with desired solubility for labeling oligonucleotides and peptides.
Catalog No. 50-195-3104 Supplier Aat Bioquest Supplier No. 2226
TQ3 is designed to be a superior quencher to TAMRA, TF3 and Cy3. TQ3 has (a). much stronger absorption; (b). much higher quenching efficiency; and (c). versatile reactive forms with desired solubility for labeling oligonucleotides and peptides.