Learn More
Selleck Chemical LLC Tideglusib S2823-200mg
SDP
Catalog No. 503109068
Encompass_Preferred
Tideglusib is an irreversible non ATP-competitive GSK-3 inhibitor with IC50 of 60 nM in a cell-free assay fails to inhibit kinases with a Cys homologous to Cys-199 located in the active site Phase 2
Catalog No. 50-310-9068 Supplier Selleck Chemical LLC Supplier No. S2823200MG
Tideglusib is an irreversible non ATP-competitive GSK-3 inhibitor with IC50 of 60 nM in a cell-free assay fails to inhibit kinases with a Cys homologous to Cys-199 located in the active site Phase 2