Learn More
Apexbio Technology LLC Tideglusib 865854-05-3 200mg
SDP
Catalog No. 5000062342
Encompass_Preferred
Tideglusib is a potent selective inhibitor of glycogen synthase kinase-3 (GSK-3 ) functioning through irreversible and ATP-noncompetitive inhibition with reported IC50 of 60 nM Mechanistically Tideglusib binds in a manner independent of ATP leading to sustained enzyme inactivation Preclinical studies utilizing animal models demonstrate Tideglusib s ability to attenuate hyperphosphorylation of Tau protein reduce amyloid plaque accumulation prevent neuronal loss and enhance cognitive performance Currently undergoing clinical investigation Tideglusib has advanced through Phase II studies for Alzheimer s disease (AD) and progressive supranuclear palsy (PSP) exploring therapeutic applicability related to its GSK-3 inhibition activity
Catalog No. 50-000-62342 Supplier Apexbio Technology LLC Supplier No. B1539200
Tideglusib is a potent selective inhibitor of glycogen synthase kinase-3 (GSK-3 ) functioning through irreversible and ATP-noncompetitive inhibition with reported IC50 of 60 nM Mechanistically Tideglusib binds in a manner independent of ATP leading to sustained enzyme inactivation Preclinical studies utilizing animal models demonstrate Tideglusib s ability to attenuate hyperphosphorylation of Tau protein reduce amyloid plaque accumulation prevent neuronal loss and enhance cognitive performance Currently undergoing clinical investigation Tideglusib has advanced through Phase II studies for Alzheimer s disease (AD) and progressive supranuclear palsy (PSP) exploring therapeutic applicability related to its GSK-3 inhibition activity