Learn More
Apexbio Technology LLC Tideglusib, 50mg. Cas: 865854-05-3 MFCD: MFCD18633296. non-ATP-competitive GSK-3β inhibitor
SDP
Supplier:  Apexbio Technology LLC B153950
Encompass_Preferred
Tideglusib (NP-12, NP031112) is a potent, selective and irreversible small molecule non-ATP-competitive GSK-3Beta inhibitor with an IC50 of 60 nM. 865854-05-3. MFCD18633296
Catalog No. 50-101-4413