Learn More
Grainger Tie Down Strap S-Hook Red
Catalog No. 502765608
Encompass_Preferred
The ANCRA 1" x 15' Tie Down Strap is a heavy duty polyester webbing strap with a working load limit of 1,200 lbs. It features double box stitching for durability, heavy duty steel S-hooks on each end, and a rubber handle grip in the center for easy handling. This cargo strap can be used for securing loads on trucks, trailers, and more. It meets DOT standards for tie downs. The strap is made in the USA.
Catalog No. 50-276-5608 Supplier Grainger Supplier No. 16V906
The ANCRA 1" x 15' Tie Down Strap is a heavy duty polyester webbing strap with a working load limit of 1,200 lbs. It features double box stitching for durability, heavy duty steel S-hooks on each end, and a rubber handle grip in the center for easy handling. This cargo strap can be used for securing loads on trucks, trailers, and more. It meets DOT standards for tie downs. The strap is made in the USA.