Learn More
Poltex Tie-On Mask Holder-2VM
SDP
Catalog No. 22000474
Change view
Click to view available options
Mounting:
Magnetic Mount
Wall Mount
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Mounting
22-000-474 Wall Mount
22-000-473 Magnetic Mount
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Consumables shown for demonstration purposes only and are not included with the product.
Catalog No. 22-000-474 Supplier Poltex Supplier No. TIEMASK2VMW
Add to Cart
Add to Cart
Consumables shown for demonstration purposes only and are not included with the product.
Visually manage two boxes of tie-on masks. One box in use, one back up.
  • When bottom box is empty, simply slide front face open, remove empty box, and full box falls into place.
  • Super durable and cleanable (alcohol resistant) PETG construction.
  • Choose from mounting methods:
    • Powerful Magnets
    • Keyhole slots (note: mounting hardware not included. Customer must provide their own screw/anchor for their wall type)
  • See image for dimensions
  • Fits 3M 1818 Tie-On Surgical Face Mask
Specifications
Product Type Face Mask Dispenser
Material PETG
For Use With (Application) Healthcare
For Use With (Equipment) Tie-on masks
Mounting Wall Mount