Learn More
Cayman Chemical TIe2 InhIbItr 7 500ug
SDP
Catalog No. 502982290
Encompass_Preferred
Blocks Tie2 kinase activity (Ki = 1.3 µM), inhibiting angiopoietin 1-induced Tie2 autophosphorylation and downstream signaling (IC50 = 0.3 µM); prevents endothelial cell tube formation and aberrant vessel growth in a rat model of Matrigel-induced choroidal neovascularization
Catalog No. 50-298-2290 Supplier Cayman Chemical Supplier No. 19535500 UG
Blocks Tie2 kinase activity (Ki = 1.3 µM), inhibiting angiopoietin 1-induced Tie2 autophosphorylation and downstream signaling (IC50 = 0.3 µM); prevents endothelial cell tube formation and aberrant vessel growth in a rat model of Matrigel-induced choroidal neovascularization