Learn More
Cayman Chemical Tie2 Inhibitor 7, Purity: ≥95%, CAS: 1020412-97-8, 5 mg
SDP
Catalog No. 502982423
Encompass_Preferred
Blocks Tie2 kinase activity (Ki = 1.3 µM), inhibiting angiopoietin 1-induced Tie2 autophosphorylation and downstream signaling (IC50 = 0.3 µM); prevents endothelial cell tube formation and aberrant vessel growth in a rat model of Matrigel-induced choroidal neovascularization
Catalog No. 50-298-2423 Supplier Cayman Chemical Supplier No. 195355 MG
Blocks Tie2 kinase activity (Ki = 1.3 µM), inhibiting angiopoietin 1-induced Tie2 autophosphorylation and downstream signaling (IC50 = 0.3 µM); prevents endothelial cell tube formation and aberrant vessel growth in a rat model of Matrigel-induced choroidal neovascularization