Learn More
TARGETMOL CHEMICALS INC Tie2 kinase inhibitor 1 5MG
SDP
Catalog No. 502877061
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Tie2 kinase inhibitor 1 (Tie2 kinase inhibitor), an optimized compound of SB-203580, is selective to Tie2 with IC50 of 0.25 uM, which is 200-fold more effective than p38. Purity 99.81%
Catalog No. 50-287-7061 Supplier TARGETMOL CHEMICALS INC Supplier No. T63355MG
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Tie2 kinase inhibitor 1 (Tie2 kinase inhibitor), an optimized compound of SB-203580, is selective to Tie2 with IC50 of 0.25 uM, which is 200-fold more effective than p38. Purity 99.81%