Learn More
Medchemexpress LLC Tie2 kinase inhibitor 1 | 948557-43-5 | 439.53 g/mol | C26H21N3O2S | 100 MG
SDP
Catalog No. 5000212687
Encompass_Preferred
Tie2 kinase inhibitor 1 is a selective small-molecule inhibitor of the Tie2 (TEK) receptor tyrosine kinase, reported to inhibit Tie2 with an IC50 of approximately 250 nM and to exhibit anti-cancer activity in preclinical studies. The compound is supplied as a solid and is intended for use in biochemical and cellular research to probe Tie2-dependent signaling pathways.
  • Potent, selective Tie2 inhibition (IC50 ≈ 250 nM).
  • Suitable for biochemical kinase assays and cellular studies.
  • Reported anti-cancer activity in preclinical models.
  • Solid form with defined storage recommendations for stability.
  • Characterized by CAS 948557-43-5 and molecular weight 439.53 g/mol.
Catalog No. 50-002-12687 Supplier Medchemexpress LLC Supplier No. HY100556100MG
Tie2 kinase inhibitor 1 is a selective small-molecule inhibitor of the Tie2 (TEK) receptor tyrosine kinase, reported to inhibit Tie2 with an IC50 of approximately 250 nM and to exhibit anti-cancer activity in preclinical studies. The compound is supplied as a solid and is intended for use in biochemical and cellular research to probe Tie2-dependent signaling pathways.
  • Potent, selective Tie2 inhibition (IC50 ≈ 250 nM).
  • Suitable for biochemical kinase assays and cellular studies.
  • Reported anti-cancer activity in preclinical models.
  • Solid form with defined storage recommendations for stability.
  • Characterized by CAS 948557-43-5 and molecular weight 439.53 g/mol.