Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tie2 kinase inhibitor 1 | 948557-43-5 | 99.8% | 439.53 | C26H21N3O2S | 1 ML
SDP

Catalog No. 5000211823
Encompass_Preferred

Tie2 kinase inhibitor 1 is a potent, selective small-molecule inhibitor of the Tie2 receptor tyrosine kinase with reported anti-cancer activity. It inhibits Tie2 with an IC50 of 250 nM and is supplied for research use.

  • Potent Tie2 inhibition with IC50 of 250 nM.
  • High purity suitable for research applications (reported 99.8%).
  • Characterized small molecule: formula C26H21N3O2S, molecular weight 439.53.
  • Physical form light yellow to yellow solid.
  • Available as concentrated solution for assay convenience.

Catalog No. 50-002-11823 Supplier Medchemexpress LLC Supplier No. HY10055610MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tie2 kinase inhibitor 1 is a potent, selective small-molecule inhibitor of the Tie2 receptor tyrosine kinase with reported anti-cancer activity. It inhibits Tie2 with an IC50 of 250 nM and is supplied for research use.

  • Potent Tie2 inhibition with IC50 of 250 nM.
  • High purity suitable for research applications (reported 99.8%).
  • Characterized small molecule: formula C26H21N3O2S, molecular weight 439.53.
  • Physical form light yellow to yellow solid.
  • Available as concentrated solution for assay convenience.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.