Learn More
Medchemexpress LLC Tie2 kinase inhibitor 1 | 948557-43-5 | MFCD18384977 | 99.8% | 439.53 g/mol | C26H21N3O2S | 25 MG
SDP
Catalog No. 5000211608
Encompass_Preferred
Tie2 kinase inhibitor 1 is a potent, selective small-molecule inhibitor of the Tie2 receptor tyrosine kinase with a reported IC50 of 250 nM and demonstrated anti-cancer activity in preclinical studies.
  • Potent Tie2 inhibition with IC50 of 250 nM.
  • High selectivity for Tie2 over related kinases.
  • Demonstrated anti-cancer activity in preclinical models.
  • High purity available (reported 99.8%).
  • Chemical formula C26H21N3O2S and molecular weight ≈ 439.53 g/mol.
  • Suitable for biochemical and cellular assays targeting Tie2 signaling.
Catalog No. 50-002-11608 Supplier Medchemexpress LLC Supplier No. HY10055625MG
Add to Cart
Add to Cart
Tie2 kinase inhibitor 1 is a potent, selective small-molecule inhibitor of the Tie2 receptor tyrosine kinase with a reported IC50 of 250 nM and demonstrated anti-cancer activity in preclinical studies.
  • Potent Tie2 inhibition with IC50 of 250 nM.
  • High selectivity for Tie2 over related kinases.
  • Demonstrated anti-cancer activity in preclinical models.
  • High purity available (reported 99.8%).
  • Chemical formula C26H21N3O2S and molecular weight ≈ 439.53 g/mol.
  • Suitable for biochemical and cellular assays targeting Tie2 signaling.