Learn More
Apexbio Technology LLC Tie2 kinase inhibitor 948557-43-5 10mM (in 1mL DMSO)
SDP
Catalog No. 5000061865
Encompass_Preferred
Tie2 kinase inhibitor (CAS 948557-43-5) is a selective and reversible small-molecule inhibitor targeting the Tie2 receptor tyrosine kinase a critical modulator of endothelial cell survival angiogenesis and vascular integrity By inhibiting Tie2 autophosphorylation this compound disrupts subsequent signaling pathways involved in endothelial maturation and tumor-associated angiogenesis In vitro studies demonstrate potent inhibition of Tie2 kinase activity (IC50 0 25 M) with substantial selectivity over other kinases including p38 In vivo treatment reduces angiogenesis and tumor progression in murine xenograft tumor models This inhibitor is thus relevant for research examining angiogenic mechanisms and cancer progression
Catalog No. 50-000-61865 Supplier Apexbio Technology LLC Supplier No. A59795.1
Tie2 kinase inhibitor (CAS 948557-43-5) is a selective and reversible small-molecule inhibitor targeting the Tie2 receptor tyrosine kinase a critical modulator of endothelial cell survival angiogenesis and vascular integrity By inhibiting Tie2 autophosphorylation this compound disrupts subsequent signaling pathways involved in endothelial maturation and tumor-associated angiogenesis In vitro studies demonstrate potent inhibition of Tie2 kinase activity (IC50 0 25 M) with substantial selectivity over other kinases including p38 In vivo treatment reduces angiogenesis and tumor progression in murine xenograft tumor models This inhibitor is thus relevant for research examining angiogenic mechanisms and cancer progression