Learn More
Medchemexpress LLC Tie2 kinase inhibitor 2 | 1020412-97-8 | 99.0% | 529.63 | 1 MG
SDP
Catalog No. 5000437970
Encompass_Preferred
Tie2 kinase inhibitor 2 is a selective Tie2 kinase inhibitor, identified as compound 7. It is effective at an IC50 value of 1 μM and can be utilized in research pertaining to Tie2-mediated angiogenic disorders.
  • Selective Tie2 kinase inhibitor
  • Inhibits endothelial cell tube formation
  • Suitable for angiogenic disorders research
Catalog No. 50-004-37970 Supplier Medchemexpress LLC Supplier No. HY1135711MG
Tie2 kinase inhibitor 2 is a selective Tie2 kinase inhibitor, identified as compound 7. It is effective at an IC50 value of 1 μM and can be utilized in research pertaining to Tie2-mediated angiogenic disorders.
  • Selective Tie2 kinase inhibitor
  • Inhibits endothelial cell tube formation
  • Suitable for angiogenic disorders research