Learn More
Selleck Chemical LLC Tie2 kinase inhibitor 50mg 948557-43-5
SDP
Catalog No. 505953
Encompass_Preferred
Tie2 kinase inhibitor is an optimized compound of SB-203580, selective to Tie2 with IC50 of 0.25 ?M, 200-fold more potent than p38. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-595-3 Supplier Selleck Chemical LLC Supplier No. S157750MG
Add to Cart
Add to Cart
Tie2 kinase inhibitor is an optimized compound of SB-203580, selective to Tie2 with IC50 of 0.25 ?M, 200-fold more potent than p38. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.