Learn More
Abcam Tie2 kinase inhibitor, ATP-binding site-targeting Tie2 kinase inhibitor. Cell-permeable., 5MG
Catalog No. 502706734
Encompass_Preferred
MW 439.5 Da, Purity >99%. Potent, reversible and selective ATP-binding site-targeting Tie2 kinase inhibitor (IC₅₀ = 250 nM). Anti- angiogenesis agent. Active in vivo and in vivo. Cell-permeable.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6734 Supplier Abcam Supplier No. AB1412705MG
MW 439.5 Da, Purity >99%. Potent, reversible and selective ATP-binding site-targeting Tie2 kinase inhibitor (IC₅₀ = 250 nM). Anti- angiogenesis agent. Active in vivo and in vivo. Cell-permeable.
The product is subject to the following: Abcam Restricted Use Statement