Learn More
Cayman Chemical TIe2 KInase InhIbItr 10mg
SDP
Catalog No. 502967955
Encompass_Preferred
Reversibly and selectively blocks Tie2 kinase activity (IC50 = 250 nM); reduces angiogenesis in a Matrigel neovascularization assay and delays tumor growth in MOPC-315 plasmacytoma and SVR angiosarcoma xenograft models
Catalog No. 50-296-7955 Supplier Cayman Chemical Supplier No. 1785810 MG
Reversibly and selectively blocks Tie2 kinase activity (IC50 = 250 nM); reduces angiogenesis in a Matrigel neovascularization assay and delays tumor growth in MOPC-315 plasmacytoma and SVR angiosarcoma xenograft models