Learn More
Cayman Chemical TIe2 KInase InhIbItr 500ug
SDP
Catalog No. 502966340
Encompass_Preferred
Reversibly and selectively blocks Tie2 kinase activity (IC50 = 250 nM); reduces angiogenesis in a Matrigel neovascularization assay and delays tumor growth in MOPC-315 plasmacytoma and SVR angiosarcoma xenograft models
Catalog No. 50-296-6340 Supplier Cayman Chemical Supplier No. 17858500 UG
Reversibly and selectively blocks Tie2 kinase activity (IC50 = 250 nM); reduces angiogenesis in a Matrigel neovascularization assay and delays tumor growth in MOPC-315 plasmacytoma and SVR angiosarcoma xenograft models