Learn More
Sino Biological Recombinant Human TIE-2 Protein (ECD, His Tag), Amine-Labeled Biotinylated
Supplier:  Sino Biological 10700H08HB100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met1-Lys745) of human Tie2 (AAA61139.1) precursor was fused with a polyhistidine tag at the C-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-211-6185