Learn More
Carnabio Usa Inc TIE2(TEK)/10UG/CYTOPLASMIC
Supplier:  Carnabio Usa Inc 0818510
Encompass
TIE2(TEK), Cytoplasmic domain, Wild type, Amino Acid 771-1124, NP_000450.1, Expressed in Insect (sf21); 10microg
Catalog No. 50-754-1005