Learn More
Eurofins Discoverx Tie2 (Y1108F), active, 10 µg. (14-766)
Supplier:  Eurofins Discoverx 14766
Encompass
N-terminal 6xHis-tagged recombinant, human Tie2 amino acids 771-end. Tyrosine 1108 has been identified as an autophosphorylation site in human Tie2. The Y1106F substitution in murine Tie2 (analogous to Y1108F in human Tie2) has been shown to block cell migration by disrupting binding of Tie2 to the docking protein Dok-R (Jones N., et al., Mol Cell Biol.(2003);23:2658-2668 and Murray B.W., et al., Biochemistry.(2001);40:10243-10253). For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9311