Learn More
Eurofins Discoverx Tie2 (Y897S), active, 10 µg. (14-764)
Supplier:  Eurofins Discoverx 14764
Encompass
N-terminal 6xHis-tagged recombinant, human Tie2 amino acids 771-end. The Y897S substitution is a gain-of-function mutation associated with hereditary venous malformations (Calvert et al., (1999), Hum. Mol. Genet. 8, 1279-1289). For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9307