Learn More
Cayman Chemical TIe2CD202b EXrAClular Dom
SDP
Catalog No. 502947726
Encompass_Preferred
Source: Active recombinant human C-terminal IgG1 Fc-His-tagged Tie2/CD202b expressed in HEK293 cells • Amino acids: 23-745 • MW: 108.5 kDa
Catalog No. 50-294-7726 Supplier Cayman Chemical Supplier No. 33988200 UG
Source: Active recombinant human C-terminal IgG1 Fc-His-tagged Tie2/CD202b expressed in HEK293 cells • Amino acids: 23-745 • MW: 108.5 kDa