Learn More
MSC Lift-All Ratchet Strap, Polyester, 2 in, 27 ft, 1,600 lb Load Capacity, 1 ea
Catalog No. 5000515605
Encompass
This polyester ratchet strap tie down is designed for secure cargo loading and storage applications. Featuring a durable 2 in. wide webbing and a yellow finish, it provides reliable strength for heavy-duty tie-down tasks.
  • Breaking strength of 5,000 lbs.
  • Working load limit of 1,600 lbs.
  • Length of 27 ft.
  • Constructed from high-strength polyester material
  • Equipped with steel flat hooks
  • Solid ratchet mandrel resists collapsing under load
  • Steel web wrap included to protect webbing from corner abrasion
  • Environmentally friendly clear zinc plating on hooks helps prevent corrosion
Catalog No. 50-005-15605 Supplier MSC Supplier No. 89751929
This polyester ratchet strap tie down is designed for secure cargo loading and storage applications. Featuring a durable 2 in. wide webbing and a yellow finish, it provides reliable strength for heavy-duty tie-down tasks.
  • Breaking strength of 5,000 lbs.
  • Working load limit of 1,600 lbs.
  • Length of 27 ft.
  • Constructed from high-strength polyester material
  • Equipped with steel flat hooks
  • Solid ratchet mandrel resists collapsing under load
  • Steel web wrap included to protect webbing from corner abrasion
  • Environmentally friendly clear zinc plating on hooks helps prevent corrosion