Learn More
Med Vet International Tiegland Modified Culture Swab, J0274, 25 Pack
Catalog No. 5000582990
Encompass_Preferred
The original uterine culture swab. Swab is covered with outer flexible plastic tube. After the culture is taken, a scored ring 31/2" from the end allows the tube to snap off and is plugged with two included plastic caps. This allows for ease of transport and storage.
Catalog No. 50-005-82990 Supplier Med Vet International Supplier No. JOR274
Add to Cart
Add to Cart
The original uterine culture swab. Swab is covered with outer flexible plastic tube. After the culture is taken, a scored ring 31/2" from the end allows the tube to snap off and is plugged with two included plastic caps. This allows for ease of transport and storage.