Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tienilic acid | 40180-04-9 | MFCD00867339 | 99.7% | 331.18 | C13H8Cl2O4S | 10 MG
SDP

Catalog No. 5000298026
Encompass_Preferred

Tienilic acid is a research-grade analytical standard used as a reference compound in biochemical and pharmacological studies. It is supplied as a solid powder with high analytical purity and should be handled and stored according to supplier safety data sheet recommendations.

  • High purity: 99.7%.
  • Molecular weight 331.18.
  • Chemical formula C13H8Cl2O4S.
  • CAS number 40180-04-9.
  • Solid form; store at 4°C protected from light.
  • In-solution storage: -80°C up to 6 months, -20°C up to 1 month.

Catalog No. 50-002-98026 Supplier Medchemexpress LLC Supplier No. HY2106510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tienilic acid is a research-grade analytical standard used as a reference compound in biochemical and pharmacological studies. It is supplied as a solid powder with high analytical purity and should be handled and stored according to supplier safety data sheet recommendations.

  • High purity: 99.7%.
  • Molecular weight 331.18.
  • Chemical formula C13H8Cl2O4S.
  • CAS number 40180-04-9.
  • Solid form; store at 4°C protected from light.
  • In-solution storage: -80°C up to 6 months, -20°C up to 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.