Learn More
Medchemexpress LLC HY-119322 10mg Medchemexpress, Tifenazoxide CAS:279215-43-9 Purity:>98%
SDP
Catalog No. 501969526
Encompass_Preferred
Medchemexpress, HY-119322 10mg Tifenazoxide CAS:279215-43-9 Tifenazoxide (NN414) is a potent, orally active and SUR1/Kir6.2 selective KATP channels opener. Tifenazoxide has antidiabetic effect, can inhibit glucose stimulated insulin release in vitro and in vivo, and has a beneficial effect on glucose homeostasis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9526 Supplier Medchemexpress LLC Supplier No. HY11932210MG
Medchemexpress, HY-119322 10mg Tifenazoxide CAS:279215-43-9 Tifenazoxide (NN414) is a potent, orally active and SUR1/Kir6.2 selective KATP channels opener. Tifenazoxide has antidiabetic effect, can inhibit glucose stimulated insulin release in vitro and in vivo, and has a beneficial effect on glucose homeostasis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.