Learn More
Medchemexpress LLC HY-119322 5mg Medchemexpress, Tifenazoxide CAS:279215-43-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1193225MG
Encompass_Preferred
Medchemexpress, HY-119322 5mg Tifenazoxide CAS:279215-43-9 Tifenazoxide (NN414) is a potent, orally active and SUR1/Kir6.2 selective KATP channels opener. Tifenazoxide has antidiabetic effect, can inhibit glucose stimulated insulin release in vitro and in vivo, and has a beneficial effect on glucose homeostasis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9527