Learn More
Aviva Systems Biology Corp TIGD1 Antibody - middle region (ARP52855_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52855_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TIGD1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TIGD1. Alternate names: EEYOREApplicationWB. Species reactivity: Dog, Horse, Human
Catalog No. 50-014-60853