Learn More
Aviva Systems Biology Corp Tigd4 Antibody - N-terminal region (ARP52858_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52858_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Tigd4. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: C130063O11Rik, Gm418ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-285-776