Learn More
Medchemexpress LLC Tigecycline | 220620-09-7 | 99.95% | 200 MG
SDP
Catalog No. 5000450502
Encompass_Preferred
Tigecycline (GAR-936) is a broad-spectrum glycylcycline antibiotic. It has been shown to inhibit AML2 and HL-60 cells, with specific IC50 values. Its activity extends to bacterial strains, demonstrating a mean inhibitory concentration (MIC) of approximately 125 ng/mL for E. coli (MG1655 strain), and MIC50 and MIC90 values of 1 and 2 mg/L, respectively, for Acinetobacter baumannii.
  • Broad-spectrum glycylcycline antibiotic
  • Inhibits AML2 and HL-60 cells in vitro
  • Demonstrates activity against E. coli and Acinetobacter baumannii
  • Light yellow to orange solid appearance
  • Derived from microorganisms
  • Reduced tumor volume and weight in an AML xenograft model
Catalog No. 50-004-50502 Supplier Medchemexpress LLC Supplier No. HYB0117200MG
Tigecycline (GAR-936) is a broad-spectrum glycylcycline antibiotic. It has been shown to inhibit AML2 and HL-60 cells, with specific IC50 values. Its activity extends to bacterial strains, demonstrating a mean inhibitory concentration (MIC) of approximately 125 ng/mL for E. coli (MG1655 strain), and MIC50 and MIC90 values of 1 and 2 mg/L, respectively, for Acinetobacter baumannii.
  • Broad-spectrum glycylcycline antibiotic
  • Inhibits AML2 and HL-60 cells in vitro
  • Demonstrates activity against E. coli and Acinetobacter baumannii
  • Light yellow to orange solid appearance
  • Derived from microorganisms
  • Reduced tumor volume and weight in an AML xenograft model