Learn More
Selleck Chemical LLC Tigecycline S1403-200mg
SDP
Catalog No. 503108340
Encompass_Preferred
Tigecycline is bacteriostatic and is a protein synthesis inhibitor by binding to the 30S ribosomal subunit of bacteria and thereby blocking entry of Aminoacyl-tRNA into the A site of the ribosome during prokaryotic translation Tigecycline induces autophagy by downregulating the PI3K-AKT-mTOR pathway
Catalog No. 50-310-8340 Supplier Selleck Chemical LLC Supplier No. S1403200MG
Tigecycline is bacteriostatic and is a protein synthesis inhibitor by binding to the 30S ribosomal subunit of bacteria and thereby blocking entry of Aminoacyl-tRNA into the A site of the ribosome during prokaryotic translation Tigecycline induces autophagy by downregulating the PI3K-AKT-mTOR pathway