Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Tigecycline

Catalog No. AC463060010
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC463060010 1 g
AC463062500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC463060010 Supplier Thermo Scientific Chemicals Supplier No. 463060010
  • $843.00
    $677.65 / Each of 1

    Save $165.35

    20% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 220620-09-7 | C29H39N5O8 | 585.66 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 220620-09-7
Molecular Formula C29H39N5O8
Molecular Weight (g/mol) 585.66
MDL Number MFCD00935753
InChI Key ZXGBRIBPJBHLMO-SBMFAFPZSA-N
IUPAC Name N-[(5aR,6aS,7S,9Z,10aS)-9-[amino(hydroxy)methylidene]-4,7-bis(dimethylamino)-1,10a,12-trihydroxy-8,10,11-trioxo-5,5a,6,6a,7,8,9,10,10a,11-decahydrotetracen-2-yl]-2-(tert-butylamino)acetamide
SMILES CN(C)[C@H]1[C@@H]2C[C@@H]3CC4=C(C=C(NC(=O)CNC(C)(C)C)C(O)=C4C(O)=C3C(=O)[C@]2(O)C(=O)\C(=C(\N)O)C1=O)N(C)C

Specifications

Melting Point 165.0°C to 175.0°C
Quantity 1 g
Formula Weight 585.65
Chemical Name or Material Tigecycline
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement May cause an allergic skin reaction.

Causes serious eye irritation.

May damage fertility or the unborn child.

Precautionary Statement GHS P Statement IF ON SKIN: Wash with plenty of soap and water.

Obtain special instructions before use.

Wear protective gloves/protective clothing/eye protection/face protection.

IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.