Learn More
Medchemexpress LLC Tigecycline-d9 | 2699607-86-6 | >98.2% | 1 MG
SDP
Catalog No. 5000450451
Encompass_Preferred
Tigecycline-d9 is a deuterium labeled form of Tigecycline, a broad-spectrum glycylcycline antibiotic. Tigecycline has a mean inhibitory concentration (MIC) for E. coli (MG1655 strain) of approximately 125 ng/mL. For Acinetobacter baumannii, the MIC50 and MIC90 are 1 and 2 mg/L, respectively. Stable heavy isotopes, such as deuterium, are incorporated into drug molecules as tracers during drug development and can influence the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled compound
  • Broad-spectrum glycylcycline antibiotic
  • Used as a tracer for quantitation during drug development
  • Potential to influence pharmacokinetic and metabolic profiles
Catalog No. 50-004-50451 Supplier Medchemexpress LLC Supplier No. HYB0117S1MG
Tigecycline-d9 is a deuterium labeled form of Tigecycline, a broad-spectrum glycylcycline antibiotic. Tigecycline has a mean inhibitory concentration (MIC) for E. coli (MG1655 strain) of approximately 125 ng/mL. For Acinetobacter baumannii, the MIC50 and MIC90 are 1 and 2 mg/L, respectively. Stable heavy isotopes, such as deuterium, are incorporated into drug molecules as tracers during drug development and can influence the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled compound
  • Broad-spectrum glycylcycline antibiotic
  • Used as a tracer for quantitation during drug development
  • Potential to influence pharmacokinetic and metabolic profiles