Learn More
Selleck Chemical LLC Tigecycline 10mg 220620-09-7 GAR-936
SDP
Catalog No. 507510
Encompass_Preferred
Tigecycline (GAR-936) is bacteriostatic and is a protein synthesis inhibitor by binding to the 30S ribosomal subunit of bacteria and thereby blocking entry of Aminoacyl-tRNA into the A site of the ribosome during prokaryotic translation. Tigecycline induces autophagy by downregulating the PI3K-AKT-mTOR pathway. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-751-0 Supplier Selleck Chemical LLC Supplier No. S140310MG
Tigecycline (GAR-936) is bacteriostatic and is a protein synthesis inhibitor by binding to the 30S ribosomal subunit of bacteria and thereby blocking entry of Aminoacyl-tRNA into the A site of the ribosome during prokaryotic translation. Tigecycline induces autophagy by downregulating the PI3K-AKT-mTOR pathway. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.